missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA3 Antibody, ProSci
Description
BOLA3 Polyclonal Antibody specifically detects DPYD in Mouse, Rat Samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | BOLA3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | WB: 1:500 - 1:2000 |
| Formulation | PBS with 0.02% sodium azide, 50% glycerol, pH7.3. |
| Gene Alias | MMDS2, bolA-like protein 3, bolA homolog 3 |
| Host Species | Rabbit |
| Purification Method | Affinity purification |
| Quantity | 0.1 mL |
| Show More |