missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ FL Iodoacetamide (BODIPY™ FL C1-IA, N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-yl)Methyl)Iodoacetamide)

Catalog No. d6003
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D6003 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D6003 Supplier Invitrogen™ Supplier No. D6003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BODIPY™ FL Iodoacetamide (BODIPY™ FL C1-IA, N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-yl)Methyl)Iodoacetamide)

The thiol-reactive BODIPY™ FL iodoacetamide can be used to create green-fluorescent bioconjugates. The electronically neutral BODIPY™ FL dye has the most fluorescein-like spectra of the green-fluorescent BODIPY™ dyes.

Specifications

Quantity 5 mg
Product Type FL Iodoacetamide
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Alkyl Halide, Iodoacetamide
Label or Dye BODIPY™ FL
Label Type BODIPY Dyes
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.