missing translation for 'onlineSavingsMsg'
Learn More

BOC-Sarcosine, 99+%

Catalog No. AC275670050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275670050 5 g Glass Bottle
AC275670250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC275670050 Supplier Thermo Scientific Chemicals Supplier No. 275670050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13734-36-6 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13734-36-6
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037795
InChI Key YRXIMPFOTQVOHG-UHFFFAOYSA-N
Synonym boc-sar-oh, boc-sarcosine, n-boc-sarcosine, glycine, n-1,1-dimethylethoxy carbonyl-n-methyl, n-tert-butoxycarbonyl-n-methylglycine, t-boc-sarcosine, n-t-boc-sarcosine, 2-tert-butoxy carbonyl methyl amino acetic acid, n-tert-butoxycarbonylsarcosine, tert-butoxycarbonyl methyl amino acetic acid
PubChem CID 83692
IUPAC Name 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)N(C)CC(=O)O

Specifications

Melting Point 86°C to 90°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass Bottle
Linear Formula (CH3)3COCON(CH3)CH2COOH
Quantity 5 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 189.21
Percent Purity ≥99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material BOC-Sarcosine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 237-306-0
TSCA TSCA

RUO – Research Use Only