missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Serine benzyl ester, 98%

Catalog No. AC434840050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC434840050 5 g
AC434840010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434840050 Supplier Thermo Scientific Chemicals Supplier No. 434840050

CAS: 59524-02-6 | C15H21NO5 | 295.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59524-02-6
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.33
InChI Key BQADRZHPZVQGCW-LBPRGKRZSA-N
Synonym boc-ser-obzl, boc-l-serine benzyl ester, s-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, l-serine, n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester, benzyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, ambotzbaa1202, pubchem12169, n-boc-serine benzyl ester, n-boc-l-serine benzyl ester
PubChem CID 7018792
IUPAC Name benzyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 67°C to 72°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Specific Rotation -14° to -18° (20°C, 589nm) (c=1, ethanol)
Formula Weight 295.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material BOC-L-Serine benzyl ester
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only