accessibility menu, dialog, popup

UserName

CAS RN 59524-02-6

H3CCH3CH3OONHHOOO

CAS RN 59524-02-6

IUPAC Name: benzyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoate
Synonyms: benzyl 2-[(tert-butoxycarbonyl)amino]-3-hydroxypropanoate
Molecular Weight (g/mol): 295.34
Molecular Formula: C15H21NO5
InChi Key: BQADRZHPZVQGCW-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)