missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BNIP1 Recombinant Rabbit Monoclonal Antibody (23GB4870)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552763
Description
BNIP1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
NIP1 (BNIP1) is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. Alternative splicing of this gene results in four products of unknown function. Transcript variant BNIP1 contains the entire coding region of the gene. This variant contains a fully conserved BH3 domain, which has been associated with pro-apoptotic function.Chemical Identifiers
| BNIP1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| BNIP1 | |
| 2010005M06Rik; 5930429G21Rik; BCL2 interacting protein 1; BCL2/adenovirus E1B 19 kDa protein-interacting protein 1; BCL2/adenovirus E1B 19kD interacting protein 1; BCL2/adenovirus E1B 19kDa interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1; BCL2/adenovirus E1B interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1-like 1; bnip1; bnip1 {ECO:0000312; Bnip1l1; EMBL:AAL50991.1, ECO:0000312; NIP1; RGD:620799}; Sec20; Sec20l; SNARE protein SEC20; transformation-related gene 8 protein; TRG8; TRG-8; Unknown (protein for MGC:133975); vesicle transport protein SEC20 | |
| BNIP1 | |
| Primary | |
| Affinity chromatography |
| 23GB4870 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q12981, Q6QD59, Q8VHI8 | |
| 140932, 224630, 662 | |
| A synthesized peptide derived from human BNIP1 (100-150AA). | |
| Antibody |
Specifications
| BNIP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BNIP1 | |
| 2010005M06Rik; 5930429G21Rik; BCL2 interacting protein 1; BCL2/adenovirus E1B 19 kDa protein-interacting protein 1; BCL2/adenovirus E1B 19kD interacting protein 1; BCL2/adenovirus E1B 19kDa interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1; BCL2/adenovirus E1B interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1-like 1; bnip1; bnip1 {ECO:0000312; Bnip1l1; EMBL:AAL50991.1, ECO:0000312; NIP1; RGD:620799}; Sec20; Sec20l; SNARE protein SEC20; transformation-related gene 8 protein; TRG8; TRG-8; Unknown (protein for MGC:133975); vesicle transport protein SEC20 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140932, 224630, 662 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4870 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q12981, Q6QD59, Q8VHI8 | |
| BNIP1 | |
| A synthesized peptide derived from human BNIP1 (100-150AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |