missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150106
Description
BMP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a protein that is capable of inducing formation of cartilage in vivo. Although other bone morphogenetic proteins are members of the TGF-beta superfamily, this gene encodes a protein that is not closely related to other known growth factors. This gene is expressed as alternatively spliced variants that share an N-terminal protease domain but differ in their C-terminal region.Specifications
| BMP1 | |
| Polyclonal | |
| Unconjugated | |
| BMP1 | |
| BMP; Bmp1; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; mammalian tolloid protein; mTld; OI13; PCOLC; Pcp; PCP2; pCP-2; procollagen C-endopeptidase; procollagen C-proteinase; procollagen C-proteinase 3; TLD | |
| Rabbit | |
| Protein A | |
| RUO | |
| 12153, 649, 83470 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P13497, P98063 | |
| BMP1 | |
| Recombinant Bone Morphogenetic Protein 1 (BMP1) (Gly856-Lys982). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |