missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BMP-4-Cell Lysates ELISA Kit

ELISA

$890.00

Specifications

Accession Number P12644
Assay Range 8.23 to 6000 pg/mL
Assay Sensitivity 15 pg/mL
Calibration Range 15 pg/mL to 6000 pg/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EHBMP4CL
Encompass
View Documents Promotion Details
Invitrogen™
EHBMP4CL
96 Tests
Pack for $890.00
Only null left
 
Description

Description

The Human Bone Morphogenic Protein 4 (Hu BMP-4) Cell lysate ELISA quantitates Hu BMP-4 in human cell and tissue lysates. The assay will exclusively recognize both natural and recombinant Hu BMP-4. Principle of the method The Human BMP-4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
Specifications

Specifications

P12644
15 pg/mL
Solid-phase sandwich ELISA
ELISA
Cell Lysates
Human
Colorimetric Microplate Reader
BMP2B,BMP2B1,MCOPS6,OFC11,ZYME
<10
HRP
Angiogenesis
2°C to 8°C
BMP4
4 hrs 45 mins
8.23 to 6000 pg/mL
15 pg/mL to 6000 pg/mL
HRP
Growth Factors & Receptors
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, BMP-6, BMP-7, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
652
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Cell lysis buffer, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Cell Lysate 20 μL
BMP2B, BMP2B1, MCOPS6, OFC11, ZYME
1 hrs 20 mins
Videos
SDS
Documents

Documents