missing translation for 'onlineSavingsMsg'
Learn More

Blue Tetrazolium 95.0+%, TCI America™

Catalog No. D09431G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D09431G Supplier TCI America Supplier No. D09431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1871-22-3
Molecular Formula C40H36Cl2N8O2
Molecular Weight (g/mol) 731.68
MDL Number MFCD00040933
InChI Key RCEHREKDVGHYAM-UHFFFAOYSA-N
Synonym blue tetrazolium chloride, tetrazolium blue, tetrazolium blue chloride, blue tetrazolium, dimethoxy neotetrazolium, 4-anisyltetrazolium blue, 3,3'-3,3'-dimethoxy-1,1'-biphenyl-4,4'-diyl bis 2,5-diphenyl-2h-tetrazol-3-ium chloride, 3,3'-dianisole-4,4'-bis 3,5-diphenyltetrazolium chloride, 3,3'-dianisolebis 4,4'-3,5-diphenyl tetrazolium chloride, 3,3'-3,3'-dimethoxy-4,4'-biphenylene bis 2,5-diphenyl-2h-tetrazolium chloride
PubChem CID 9853362
ChEBI CHEBI:75198
IUPAC Name dihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1

Specifications

Melting Point 248°C
Color Yellow
Quantity 1 g
Formula Weight 727.65
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Blue Tetrazolium