missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blue Tetrazolium 95.0+%, TCI America™
$74.56 - $223.68
Chemical Identifiers
| CAS | 1871-22-3 |
|---|---|
| Molecular Formula | C40H36Cl2N8O2 |
| Molecular Weight (g/mol) | 731.68 |
| MDL Number | MFCD00040933 |
| InChI Key | RCEHREKDVGHYAM-UHFFFAOYSA-N |
| Synonym | Ditetrazolium Chloride, Tetrazolium Blue |
| PubChem CID | 9853362 |
| ChEBI | CHEBI:75198 |
| IUPAC Name | dihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride |
| SMILES | [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1 |
Chemical Identifiers
| 1871-22-3 | |
| 731.68 | |
| RCEHREKDVGHYAM-UHFFFAOYSA-N | |
| 9853362 | |
| dihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride |
| C40H36Cl2N8O2 | |
| MFCD00040933 | |
| Ditetrazolium Chloride, Tetrazolium Blue | |
| CHEBI:75198 | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1 |
Specifications
| 248°C | |
| C40H36Cl2N8O2 | |
| Ditetrazolium Chloride, Tetrazolium Blue | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1 | |
| 731.68 | |
| CHEBI:75198 | |
| ≥95.0% (T) | |
| 1 g | |
| Blue Tetrazolium [for Biochemical Research] |
| 1871-22-3 | |
| MFCD00040933 | |
| RCEHREKDVGHYAM-UHFFFAOYSA-N | |
| dihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride | |
| 9853362 | |
| 727.65 | |
| Yellow | |
| Crystal/Powder |
Safety and Handling
TSCA : Yes