missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blue Tetrazolium 95.0+%, TCI America™
Supplier: TCI America B35815G
Specifications
| Blue Tetrazolium [for Biochemical Research] | |
| 1871-22-3 | |
| MFCD00040933 | |
| RCEHREKDVGHYAM-UHFFFAOYSA-N | |
| dihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride | |
| 9853362 | |
| 727.65 | |
| Yellow | |
| Crystal/Powder |
| 248°C | |
| C40H36Cl2N8O2 | |
| Ditetrazolium Chloride, Tetrazolium Blue | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1 | |
| 731.68 | |
| CHEBI:75198 | |
| ≥95.0% (T) | |
| 5 g |
Chemical Identifiers
| 1871-22-3 | |
| 731.68 | |
| RCEHREKDVGHYAM-UHFFFAOYSA-N | |
| 9853362 | |
| dihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride |
| C40H36Cl2N8O2 | |
| MFCD00040933 | |
| Ditetrazolium Chloride, Tetrazolium Blue | |
| CHEBI:75198 | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1 |
Safety and Handling
TSCA : Yes