Learn More
Blue Tetrazolium chloride
CAS: 1871-22-3 | C40H36Cl2N8O2 | 731.68 g/mol
Supplier: Thermo Scientific Chemicals A1250203
Description
Blue Tetrazolium chloride is useful in the prepartion of DNA cation complexes and immunoadsorbents for anti-DNA antibodies. It is used in assay of reducing sugars. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBlue Tetrazolium chloride is useful in the prepartion of DNA cation complexes and immunoadsorbents for anti-DNA antibodies. It is used in assay of reducing sugars. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum.
Solubility
Soluble in hot water (5 mg/ml), methanol (50 mg/ml), ethanol, and chloroform. Insoluble in acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| Blue Tetrazolium chloride | |
| ∼255°C (decomposition) | |
| MFCD00040933 | |
| 3894077 | |
| blue tetrazolium chloride, tetrazolium blue, tetrazolium blue chloride, blue tetrazolium, dimethoxy neotetrazolium, 4-anisyltetrazolium blue, 3,3'-3,3'-dimethoxy-1,1'-biphenyl-4,4'-diyl bis 2,5-diphenyl-2h-tetrazol-3-ium chloride, 3,3'-dianisole-4,4'-bis 3,5-diphenyltetrazolium chloride, 3,3'-dianisolebis 4,4'-3,5-diphenyl tetrazolium chloride, 3,3'-3,3'-dimethoxy-4,4'-biphenylene bis 2,5-diphenyl-2h-tetrazolium chloride | |
| RCEHREKDVGHYAM-UHFFFAOYSA-N | |
| 2-[4-[4-(3,5-diphenyltetrazol-2-ium-2-yl)-3-methoxyphenyl]-2-methoxyphenyl]-3,5-diphenyltetrazol-2-ium;dichloride | |
| 9853362 | |
| 727.66 |
| 1871-22-3 | |
| C40H36Cl2N8O2 | |
| 1 g | |
| 14,9244 | |
| Soluble in hot water (5mg/ml),methanol (50mg/ml),ethanol,and chloroform. Insoluble in acetone. | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1 | |
| 731.68 | |
| CHEBI:75198 |
Chemical Identifiers
| 1871-22-3 | |
| 731.68 | |
| RCEHREKDVGHYAM-UHFFFAOYSA-N | |
| 9853362 | |
| 2-[4-[4-(3,5-diphenyltetrazol-2-ium-2-yl)-3-methoxyphenyl]-2-methoxyphenyl]-3,5-diphenyltetrazol-2-ium;dichloride |
| C40H36Cl2N8O2 | |
| MFCD00040933 | |
| blue tetrazolium chloride, tetrazolium blue, tetrazolium blue chloride, blue tetrazolium, dimethoxy neotetrazolium, 4-anisyltetrazolium blue, 3,3'-3,3'-dimethoxy-1,1'-biphenyl-4,4'-diyl bis 2,5-diphenyl-2h-tetrazol-3-ium chloride, 3,3'-dianisole-4,4'-bis 3,5-diphenyltetrazolium chloride, 3,3'-dianisolebis 4,4'-3,5-diphenyl tetrazolium chloride, 3,3'-3,3'-dimethoxy-4,4'-biphenylene bis 2,5-diphenyl-2h-tetrazolium chloride | |
| CHEBI:75198 | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1 |
Safety and Handling
EINECSNumber : 217-488-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only