Learn More
Keystone Cleanroom Products Blue SMS Sterilization Wraps, 88 gsm
SMS sterilization wraps provide a microbial barrier and are ideal for pharmaceutical production applications. They are designed for use in steam, ethylene oxide, and hydrogen peroxide sterilization processes.
$540.85 - $2,747.02
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Certifications/Compliance | COC |
| Color | Blue |
| Disposable | Single-use |
| Material | SMS Fabric |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
|
17101122
|
Keystone Cleanroom Products
KCRP60001.5X01.5 |
3.8 x 3.8 cm (1.5 x 1.5 in.) |
Case of 3000 Each for $811.30
|
|
|||||
|
17101123
|
Keystone Cleanroom Products
KCRP6000202 |
76.2 cm |
Case of 3000 Each for $811.30
|
|
|||||
|
17101124
|
Keystone Cleanroom Products
KCRP6000303 |
7.6 x 7.6 cm (3 x 3 in.) |
Case of 3000 Each for $811.30
|
|
|||||
|
17101125
|
Keystone Cleanroom Products
KCRP6000404 |
10.2 x 10.2 cm (4 x 4 in.) |
Case of 3000 Each for $811.30
|
|
|||||
|
17101126
|
Keystone Cleanroom Products
KCRP6000606 |
15.2 x 15.2 cm (6 x 6 in.) |
Case of 3000 Each for $811.30
|
|
|||||
|
17101127
|
Keystone Cleanroom Products
KCRP6000808 |
20.3 x 20.3 cm (8 x 8 in.) |
Case of 1000 Each for $540.85
|
|
|||||
|
17101128
|
Keystone Cleanroom Products
KCRP6000909 |
22.9 x 22.9 cm (9 x 9 in.) |
Case of 1000 Each for $597.79
|
|
|||||
|
17101129
|
Keystone Cleanroom Products
KCRP6000912 |
22.9 x 30.5 cm (9 x 12 in.) |
Case of 1000 Each for $654.73
|
|
|||||
|
17101130
|
Keystone Cleanroom Products
KCRP6001212 |
30.5 x 30.5 cm (12 x 12 in.) |
Case of 1000 Each for $782.83
|
|
|||||
|
17101131
|
Keystone Cleanroom Products
KCRP6001515 |
38.1 x 38.1 cm (15 x 15 in.) |
Case of 1000 Each for $1,010.55
|
|
|||||
|
17101132
|
Keystone Cleanroom Products
KCRP6001818 |
45.7 x 45.7 cm (18 x 18 in.) |
Case of 1000 Each for $1,409.10
|
|
|||||
|
17101133
|
Keystone Cleanroom Products
KCRP6001822 |
45.7 x 55.9 cm (18 x 22 in.) |
Case of 1000 Each for $2,747.02
|
|
|||||
|
17101134
|
Keystone Cleanroom Products
KCRP6002020 |
50.8 x 50.8 cm (20 x 20 in.) |
Case of 1000 Each for $1,380.63
|
|
|||||
|
17101135
|
Keystone Cleanroom Products
KCRP6002424 |
61 x 61 cm (24 x 24 in.) |
Case of 250 Each for $583.56
|
|
|||||
|
17101136
|
Keystone Cleanroom Products
KCRP6003030 |
76.2 x 76.2 cm (30 x 30 in.) |
Case of 250 Each for $996.32
|
|
|||||
|
17101137
|
Keystone Cleanroom Products
KCRP6003636 |
91.4 x 91.4 cm (36 x 36 in.) |
Case of 250 Each for $1,380.63
|
|
|||||
|
17101138
|
Keystone Cleanroom Products
KCRP6004040 |
101.6 x 101.6 cm (40 x 40 in.) |
Case of 250 Each for $1,750.70
|
|
|||||
|
17101139
|
Keystone Cleanroom Products
KCRP6004545 |
114.3 x 114.3 cm (45 x 45 in.) |
Case of 250 Each for $2,120.75
|
|
|||||
|
17101140
|
Keystone Cleanroom Products
KCRP6004548 |
114.3 x 121.9 cm (45 x 48 in.) |
Case of 250 Each for $2,362.73
|
|
|||||
|
17101141
|
Keystone Cleanroom Products
KCRP6004848 |
121.9 x 121.9 cm (48 x 48 in.) |
Case of 250 Each for $2,362.73
|
|
|||||
|
17101142
|
Keystone Cleanroom Products
KCRP6005454 |
137.2 x 137.2 cm (54 x 54 in.) |
Case of 100 Each for $996.32
|
|
|||||
|
17101143
|
Keystone Cleanroom Products
KCRP6005472 |
137.2 x 182.9 cm (54 x 72 in.) |
Case of 100 Each for $1,594.13
|
|
|||||
|
17101144
|
Keystone Cleanroom Products
KCRP6006060 |
152.4 x 152.4 cm (60 x 60 in.) |
Case of 100 Each for $1,651.05
|
|
|||||
|
17101145
|
Keystone Cleanroom Products
KCRP6006080 |
152.4 x 203.2 cm (60 x 80 in.) |
Case of 50 Each for $868.22
|
|
|||||
Description
- Manufactured and packaged in a Certified ISO 8 cleanroom
- Clean - no cellulose
- Strong, rip and tear resistant 88 gsm material
- Soft and drape-able fabric
- An extensive range of sizes available means less cutting
- Hydrophobic barrier allows sterilization gasses but prevents liquid and microbial penetration
- Sub packed into sealed labeled poly bags with Lot Number and Date of Manufacture for complete traceability
- Certificate of Conformance sent with each shipment
- Made in the USA
Specifications
| Autoclavable | |
| Blue | |
| SMS Fabric | |
| Yes |
| COC | |
| Single-use | |
| Non-sterile | |
| Sterilization Wrap (Super-duty) |