missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Blue PTFE/White Silicone Septa

It is used as an air-tight seal, and prevents contamination

Supplier:  MilliporeSigma™ Supelco™ SGMA27511

 View more versions of this product

Catalog No. 111011249


Only null left

Description

Description

Specifications

Chemical Identifiers

Blue, White
PTFE/Silicone
100

Specifications

11 mm
PTFE/Silicone
Max. temp.: 250° C
4 mL Vial
Blue, White
100
0.06 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions