Learn More
Fisherbrand™ Blood Bank Saline 0.85% - 0.9%
Fisherbrand Blood Bank Saline 0.85% - 0.9% is a quality blood bank saline product that is azide- and preservative-free.
Supplier: Fisherbrand™ 293 184
Description
Special processing assures extremely low particle count when you use Fisherbrand™ Blood Bank Saline 0.85% - 0.9%, a quality blood bank saline product that is azide- and preservative-free.
- Consists of a w/v isotonic NaCl solution
- Special processing assures extremely low particle count
- For in vitro diagnostic and laboratory use only
- 2.64 gal. (10L) and 5.28 gal. (20L) sizes supplied in cubitainers
Non-Buffered Saline
- W/V: 0.9%
- pH: 6.0 – 7.5
Buffered Saline
- W/V: 0.85%
- pH: 7.2 ± 0.1
- Phosphate buffered
Note:
Not for infusion, injection or irrigation.
Specifications
| Saline, Non-buffered 0.9% | |
| 1 gal. |