missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Blood Bank Saline 0.85% - 0.9%

Catalog No. p-4527840
Change view
Click to view available options
Quantity:
1 gal.
10 L
20 L
Type:
Saline, Buffered 0.85%
Saline, Non-buffered 0.9%
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23535435 20 L Saline, Non-buffered 0.9%
23312651 10 L Saline, Buffered 0.85%
23309178 20 L Saline, Buffered 0.85%
23293184 1 gal. Saline, Non-buffered 0.9%
23062125 10 L Saline, Non-buffered 0.9%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 23535435 Supplier Fisherbrand™ Supplier No. 535 435
Only null left

Fisherbrand Blood Bank Saline 0.85% - 0.9% is a quality blood bank saline product that is azide- and preservative-free.

Special processing assures extremely low particle count when you use Fisherbrand™ Blood Bank Saline 0.85% - 0.9%, a quality blood bank saline product that is azide- and preservative-free.

  • Consists of a w/v isotonic NaCl solution
  • Special processing assures extremely low particle count
  • For in vitro diagnostic and laboratory use only
  • 2.64 gal. (10L) and 5.28 gal. (20L) sizes supplied in cubitainers

Non-Buffered Saline

  • W/V: 0.9%
  • pH: 6.0 – 7.5

Buffered Saline

  • W/V: 0.85%
  • pH: 7.2 ± 0.1
  • Phosphate buffered

Note:

Not for infusion, injection or irrigation.

Specifications

Type Saline, Non-buffered 0.9%
Quantity 20 L