missing translation for 'onlineSavingsMsg'
Learn More

Corepoint Scientific™ Blood Bank Refrigerator
GREENER_CHOICE

This Blood Bank Refrigerator is engineered to provide quality storage and maximum protection for life-saving blood. In compliance with the requirements set forth by AABB for the refrigerated cold storage of blood-based products and in accordance with FDA listed Class II medical devices.

Supplier:  Corepoint Scientific™ NSBR492WSG/0

 View more versions of this product

Catalog No. 22651393

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Optimal temperature control with PID microprocessor
  • Smart controller
  • EPA SNAP compliant refrigerants
  • Full array of alarms
  • LED interior lighting
  • Stainless steel interiors
  • Sliding drawers
  • Probe access port
Specifications

Chemical Identifiers

Blood Refrigerator
PID Microprocessor
Painted Steel
Urethane
Upright
White
Probe Access Port
Integrated Alarm Systems
NEMA 5-15
1°C to 10°C
Natural Hydrocarbon Refrigerant
Powder-coated Steel
EPA SNAP, FDA, AABB
LCD Display
Fan-forced
Cycle
V-drive
100/120 V

Specifications

Blood Refrigerator
1388 L
PID Microprocessor
Swing Glass
7 Drawers per Side (14 Total), Optional Extra Drawer per Side (16 Total)
Powder-coated Steel
EPA SNAP, FDA, AABB
LCD Display
Fan-forced
2 Year Warranty for Parts and Labor (Excluding Calibrations), 7 Year Warranty for Compressor
Integrated Alarm Systems
4.5 A
100/120 V
49 cu. ft.
1°C to 10°C
Natural Hydrocarbon Refrigerant
2
Painted Steel
Urethane
Upright
White
Probe Access Port
Cycle
V-drive
NEMA 5-15
Blood Bags (800+)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions