missing translation for 'onlineSavingsMsg'
Learn More

Blonanserin 98.0+%, TCI America™

Supplier:  TCI America B456525MG

 View more versions of this product

Catalog No. B456525MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Blonanserin
124°C
C23H30FN3
25 mg
XVGOZDAJGBALKS-UHFFFAOYSA-N
1-ethyl-4-[4-(4-fluorophenyl)-5H,6H,7H,8H,9H,10H-cycloocta[b]pyridin-2-yl]piperazine
125564
≥98.0% (HPLC)
132810-10-7
White
MFCD00893838
2-(4-Ethyl-1-piperazinyl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine
CCN1CCN(CC1)C1=CC(C2=CC=C(F)C=C2)=C2CCCCCCC2=N1
367.51
367.51
Crystalline Powder

Chemical Identifiers

132810-10-7
367.51
XVGOZDAJGBALKS-UHFFFAOYSA-N
125564
CCN1CCN(CC1)C1=CC(C2=CC=C(F)C=C2)=C2CCCCCCC2=N1
C23H30FN3
MFCD00893838
2-(4-Ethyl-1-piperazinyl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine
1-ethyl-4-[4-(4-fluorophenyl)-5H,6H,7H,8H,9H,10H-cycloocta[b]pyridin-2-yl]piperazine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GY0175140

TSCA : No

SDS
Documents

Documents

Promotions

Promotions