missing translation for 'onlineSavingsMsg'
Learn More

Blonanserin 98.0+%, TCI America™

Catalog No. p-7123621
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B456525MG 25 mg
B4565100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B456525MG Supplier TCI America Supplier No. B456525MG
Only null left

Chemical Identifiers

CAS 132810-10-7
Molecular Formula C23H30FN3
Molecular Weight (g/mol) 367.51
MDL Number MFCD00893838
InChI Key XVGOZDAJGBALKS-UHFFFAOYSA-N
Synonym 2-(4-Ethyl-1-piperazinyl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine
PubChem CID 125564
IUPAC Name 1-ethyl-4-[4-(4-fluorophenyl)-5H,6H,7H,8H,9H,10H-cycloocta[b]pyridin-2-yl]piperazine
SMILES CCN1CCN(CC1)C1=CC(C2=CC=C(F)C=C2)=C2CCCCCCC2=N1

Specifications

Melting Point 124°C
Color White
Quantity 25 mg
Formula Weight 367.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Blonanserin
RTECSNumber GY0175140
TSCA No