missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ FL Fluorescent Blocking Buffer (10X)

Catalog No. PI37565
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI37565 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI37565 Supplier Thermo Scientific™ Supplier No. 37565

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Blocker FL Fluorescent Blocking Buffer is designed to block excess nonspecific binding sites to help reduce background fluorescence in western blotting applications.

Blocker FL buffer helps enhance fluorescent protein detection by increasing the signal-to-noise ratio in blotting applications where background fluorescence may otherwise interfere with signal detection.

Compare and view all available blocking buffers

Working reagent is prepared by adding 1 part Blocker FL buffer to 9 parts ultrapure water. Optimal concentration should be determined empirically for each system. Compatible with both nitrocellulose and PVDF membranes.

Specifications

Chemical Name or Material Blocker FL
Concentration 10X
For Use With (Application) Western Blot
Physical Form Liquid
Product Line Blocker
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.