missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ Casein

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.

Supplier:  Thermo Scientific™ LSG37532

 View more versions of this product

Catalog No. PI37532


Only null left

Description

Description

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Features of Blocker Casein include:

  • Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
  • Easy to use—1% casein solution is ready to use; can be diluted further as needed
  • Safe—stable, thimerosal-free formulation
Specifications

Chemical Identifiers

1X
Western Blot
Liquid
Blocker

Specifications

TBS
Western Blot
Blocker
1X
Liquid
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C.

SDS
Documents

Documents

Promotions

Promotions