missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ Casein

Catalog No. FER37528X3
Change view
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L
Chemical Name or Material:
PBS
TBS
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Chemical Name or Material
FER37528X3 3 x 1 L PBS
PI37582 100 mL PBS
PI37528 1 L PBS
PI37583 100 mL TBS
PI37532 1 L TBS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. FER37528X3 Supplier Thermo Scientific™ Supplier No. 37528X3

missing translation for 'validMainframeMsg'

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Features of Blocker Casein include:

  • Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
  • Easy to use—1% casein solution is ready to use; can be diluted further as needed
  • Safe—stable, thimerosal-free formulation

Specifications

Chemical Name or Material PBS
Concentration 1X
For Use With (Application) Western Blot
Physical Form Liquid
Product Line Blocker
Quantity 3 x 1 L
What is the main ingredient in Blocker Casein (Cat. No. 37582, 37528, 37528X3)?

Blocker Casein (Cat. No. 37582, 37528, 37528X3) is a ready-to-use, 1% (w/v) solution of purified casein in phosphate-buffered saline (PBS). It is a stable, thimerosal-free formulation and is already at the optimal concentration for most applications. However, it can be diluted further if needed.