missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Blocker™ BSA
Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Supplier: Thermo Scientific™ 37520
Description
Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Blocker BSA is a 10% (w/v) solution of high-quality BSA that is useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. It is most frequently diluted 10-fold (to 1% BSA) in 1X PBS or 1X TBS for initial testing. Blocker BSA is usually more effective than nonfat milk for biotin-avidin systems because it contains a single purified protein that is devoid of endogenous biotin.Chemical Identifiers
| 10X | |
| Liquid | |
| 10% BSA |
| Western Blot | |
| Blocker |
Specifications
| TBS | |
| 10X | |
| Liquid | |
| 125 mL |
| Blocker BSA in TBS (10X) | |
| Western Blot | |
| Blocker | |
| 10% BSA |
Safety and Handling
Recommended Storage : Store at 4°C.