missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BLOCK-iT™ Transfection Optimization Kit

Catalog No. 13750047
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13750047 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13750047 Supplier Invitrogen™ Supplier No. 13750047

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BLOCK-iT™ Transfection Optimization Kit

The BLOCK-iT™ Transfection Optimization Kit is designed to optimize RNAi transfection using controls for transfection and viability. Each kit contains a BLOCK-iT™ Fluorescent Oligo, which provides:

  • A strong, easily detectable FITC signal indicating transfection efficiency
  • Chemical modifications for a clearer, more persistent cellular signal than labeled siRNA provides
  • Proven correlation of transfection efficiency with Stealth RNAi™ siRNA and traditional siRNA molecules

    This makes BLOCK-iT™ Fluorescent Oligo an ideal indicator of transfection efficiency for RNAi experiments. In addition, the kit contains a Positive Control Stealth RNAi™ siRNA molecule directed at human p53 and a Scrambled Stealth RNAi™ siRNA molecule as a negative control. To assay cell viability, Dead Cell Dye is included. This dye colors dead cells as red, thereby allowing easy identification of non-viable cells in a tissue culture plate. Using these reagents together allows you to both optimize transfection conditions and have a functional positive control in every RNAi experiment you perform.
  • Specifications

    Control Type Cell Viability Control, Negative Control, Transfection Control, Positive Control
    Format Kit
    Sample Type RNA
    Vector pLATE
    For Use With (Application) Transfection
    Product Line BLOCK-iT™, Stealth RNAi™ siRNA
    Product Type Transfection Kit
    Quantity 1 Kit
    RNAi Type siRNA

    For Research Use Only. Not for use in diagnostic procedures.