missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLOCK-iT™ Transfection Kit
Description
The BLOCK-iT™ Transfection Kit contains everything you need for successful high-efficiency transfection of siRNA into cells. Each kit contains Lipofectamine™ 2000 Transfection Reagent for efficient delivery in a broad range of cell lines and the BLOCK-iT™ Fluorescent Oligo for use as an indicator of transfection efficiency. The BLOCK-iT™ Fluorescent Oligo provides:
- A strong, easily detectable fluorescein signal indicating transfection efficiency
- Chemical modifications for a clearer, more persistent cellular signal than labeled siRNA
- Proven correlation of transfection efficiency with Stealth RNAi™ siRNA and traditional siRNA molecules
Specifications
Specifications
| Content And Storage | Each kit contains 125 μl of 20 μM BLOCK-iT™ Fluorescent Oligo and 250 μl of Lipofectamine™ 2000 Reagent. Store the BLOCK-iT™ Fluorescent Oligo at -20°C and Lipofectamine™ 2000 at +4°C |
| Control Type | Transfection Control |
| Format | Kit |
| Sample Type | Synthetic siRNA |
| Transfection Technique | Lipid-based Transfection |
| For Use With (Application) | Transfection |
| Product Line | BLOCK-iT |
| Product Type | Transfection Kit |
| Quantity | 1 Kit |
| Reagent Type | RNA |
For Research Use Only. Not for use in diagnostic procedures.