missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Block Heater
Heating Blocks Sold Separately
$1,524.78
Specifications
| Description | Block Not Included |
|---|---|
| Temperature Range (Metric) | Ambient to 150°C |
| For Use With (Application) | Incubation/Activation of Cultures, Enzyme Reactions, Immunoassays, Melting/Boiling Points, and More |
| Heat Up Time | 45 minutes (to 100°C) |
| Includes | Thermometer Well |
Description
- Multi-purpose unit ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures
- Accepts separate, interchangeable modular heating blocks which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes
- Two Temperature Adjustments: Ambient to 100°C (Low Range) or 75° to 150°C (High Range)
- Blocks include a thermometer well for measuring block temperature
- Economical, versatile and compact
- Heating Blocks sold separately
Specifications
| Block Not Included | |
| Incubation/Activation of Cultures, Enzyme Reactions, Immunoassays, Melting/Boiling Points, and More | |
| Thermometer Well | |
| ±0.1°C (at 37°C) | |
| 5.8 lb. | |
| 110 watts |
| Ambient to 150°C | |
| 45 minutes (to 100°C) | |
| ±0.1°C (at 37°C) | |
| 12.4 x 8 x 3.5 in. (31.5 x 20.3 x 8.9 cm) | |
| 2.6 kg |