missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Block Heater

Catalog No. 111008015
Change view
Click to view available options
Voltage:
110 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
111008015 110 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111008015 Supplier MilliporeSigma™ Supelco™ Supplier No. 33315

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Heating Blocks Sold Separately

  • Multi-purpose unit ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures
  • Accepts separate, interchangeable modular heating blocks which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes
  • Two Temperature Adjustments: Ambient to 100°C (Low Range) or 75° to 150°C (High Range)
  • Blocks include a thermometer well for measuring block temperature
  • Economical, versatile and compact
  • Heating Blocks sold separately

Specifications

Description Block Not Included
Plug Type US
Temperature Range (Metric) Ambient to 150°C
For Use With (Application) Incubation/Activation of Cultures, Enzyme Reactions, Immunoassays, Melting/Boiling Points, and More
Heat Up Time 45 minutes (to 100°C)
Includes Thermometer Well
Temperature Stability ±0.1°C (at 37°C)
Temperature Uniformity ±0.1°C (at 37°C)
Dimensions (L x W x H) 12.4 x 8 x 3.5 in. (31.5 x 20.3 x 8.9 cm)
Shipping Weight (English) 5.8 lb.
Shipping Weight (Metric) 2.6 kg
Voltage 110 V
Wattage 110 watts
Amperage 0.9 amps
Show More Show Less