missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bleomycin Sulfate (mixture) 85.0+%, TCI America™
Supplier: TCI America B397250MG
Specifications
| Bleomycin Sulfate (mixture) | |
| Crystalline Powder | |
| White/Yellow | |
| C55H85N17O25S4 | |
| WUIABRMSWOKTOF-UHFFFAOYSA-N | |
| [2-[2-[2-[[6-amino-2-[3-amino-1-[(2,3-diamino-3-oxopropyl)amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[[5-[[1-[2-[4-[4-(3-dimethylsulfoniopropylcarbamoyl)-1,3-thiazol-2-yl]-1,3-thiazol-2-yl]ethylamino]-3-hydroxy-1-oxobutan-2-yl]amino]-3-hyd | |
| 131664136 |
| Bleomycin Sulfate (mixture) | |
| 9041-93-4 | |
| 50 mg | |
| MFCD00070310 | |
| CC1=C(N=C(N=C1N)C(CC(=O)N)NCC(C(=O)N)N)C(=O)NC(C(C2=CN=CN2)OC3C(C(C(C(O3)CO)O)O)OC4C(C(C(C(O4)CO)O)OC(=N)[O-])O)C(=O)NC(C)C(C(C)C(=O)NC(C(C)O)C(=O)NCCC5=NC(=CS5)C6=NC(=CS6)C(=O)NCCC[S+](C)C)O.OS(=O)(=O)O | |
| 1512.619 | |
| ≥85.0% (HPLC) |
Chemical Identifiers
| 9041-93-4 | |
| 1512.619 | |
| WUIABRMSWOKTOF-UHFFFAOYSA-N | |
| [2-[2-[2-[[6-amino-2-[3-amino-1-[(2,3-diamino-3-oxopropyl)amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[[5-[[1-[2-[4-[4-(3-dimethylsulfoniopropylcarbamoyl)-1,3-thiazol-2-yl]-1,3-thiazol-2-yl]ethylamino]-3-hydroxy-1-oxobutan-2-yl]amino]-3-hyd |
| C55H85N17O25S4 | |
| MFCD00070310 | |
| 131664136 | |
| CC1=C(N=C(N=C1N)C(CC(=O)N)NCC(C(=O)N)N)C(=O)NC(C(C2=CN=CN2)OC3C(C(C(C(O3)CO)O)O)OC4C(C(C(C(O4)CO)O)OC(=N)[O-])O)C(=O)NC(C)C(C(C)C(=O)NC(C(C)O)C(=O)NCCC5=NC(=CS5)C6=NC(=CS6)C(=O)NCCC[S+](C)C)O.OS(=O)(=O)O |
Safety and Handling
RTECSNumber : EC5991990
TSCA : No
Recommended Storage : Refrigerator