missing translation for 'onlineSavingsMsg'
Learn More

UVP Blak-Ray™ B-100A UV Lamps

High powered lamps offer brilliant irradiance for optimum fluorescence

Supplier:  UVP 95004422

 View more versions of this product

Catalog No. UVP95004422

Only null left

Description

Description

  • Features 100 watts of 365nm longwave UV
  • Manufactured with scratch-resistant, powder painted aluminum lamp head
  • Lamp head is connected to transformer base by 2.44m (8 ft.) cord
  • Handle fits into transformer base allowing lamp head to rotate full 360°

95-0044-03:

  • Includes 20 ft. (6.1m) primary and secondary cords for additional mobility
Specifications

Chemical Identifiers

UV-A
100 W
1 Ea.
115 V
365 nm
Longwave UV Lamp

Specifications

UV-A
100 W
B100A
Longwave UV Lamp
115 V
365 nm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions