Learn More
Black Stallion™ Elkskin Leather Welder's Gloves with Nomex™ Lining
Extra thick, elkskin leather construction provides heat, flame, and abrasion resistance. DuPont™ Nomex™ liner on the back of the glove offers additional fire protection.
Supplier: Black Stallion™ R850M
Description
- Elkskin leather construction is extra thick to provide heat, flame, and abrasion resistance
- DuPont™ Nomex liner offers additional fire-resistant protection
- Aramid Fiber stitching for spark and abrasion resistance
- Reversed grain leather palm enhances grip, and provides a softer feel on the hand
- Thumb strap provides additional reinforcement or high stress areas
Specifications
| Gold | |
| Medium | |
| White | |
| Nomex™ | |
| 850 |
| Elkskin Leather | |
| Stick Welding | |
| Gauntlet | |
| Grain Leather |