missing translation for 'onlineSavingsMsg'
Learn More

Black Stallion™ Elkskin Leather Welder's Gloves with Nomex™ Lining

Catalog No. p-144329
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Catalog No. Size
189991314A Small
189991314B Medium
189991314C Large
189991314D X-Large
4 options

Catalog No. 189991314A

Supplier: Black Stallion™ 850S

Only null left

Extra thick, elkskin leather construction provides heat, flame, and abrasion resistance. DuPont™ Nomex™ liner on the back of the glove offers additional fire protection.

  • Elkskin leather construction is extra thick to provide heat, flame, and abrasion resistance
  • DuPont™ Nomex liner offers additional fire-resistant protection
  • Aramid Fiber stitching for spark and abrasion resistance
  • Reversed grain leather palm enhances grip, and provides a softer feel on the hand
  • Thumb strap provides additional reinforcement or high stress areas

Specifications

Color Gold
Material Elkskin Leather
Size Small
For Use With (Application) Stick Welding
Glove Cuff Color White
Glove Cuff Style Gauntlet
Liner Material Nomex™
Palm Type Grain Leather
Series 850