Learn More
Black Stallion™ Cape Sleeves
Offer protection even from molten metal splash
Supplier:  Black Stallion™ 65AHSXL
 View more versions of this product
Catalog No. 18-999-1513
Only null left
Explore available promotions
Alert:
Caution!Garments are not designed to offer long-term protection from ambient heat or direct flame contact. Not for fire protection.
Description
Description
  • Aluminized clothing designed to reflect radiant heat up to 1093°C (2000°F)
  • All of these garments are of 19 oz. aluminized heavy carbon-Kevlar™ blend fabric
  • Seams are sewn with Kevlar thread
Specifications
Specifications
X-Large
Silver
19 oz.
Aluminized Carbon, Dupont™ Kevlar™ aramid fibers
50 in.
538.6 g
Product Suggestions
Product Suggestions