Learn More
MilliporeSigma™ Supelco™ Black Plastic Caps
Description
- Solid top
- PTFE-faced rubber liner
- Prevents contamination
- Easy sterilization
Specifications
Specifications
| Material (Liner) | PTFE-Faced Rubber |
| Color | Black |
| Material | Phenolic |
| No. per Pack | 200/Pk. |
| Closure Size | 13-425 |
| Autoclavable | Autoclavable |
| Product Type | Solid Top Cap |