Learn More
Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps
Used for food, beverage, medical and chemical applications packed at <120°F. Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps are ideal for general use.
Supplier: Qorpak™ CAP00165
Description
- HDPE coated paper liner
- Used for mild acids, alkalis, solvents, alcohols, oils and aqueous products
- Not recommended for active hydrocarbons or bleaches
- Available in smooth and ribbed style
- Available as bulk/bagged packaging
Chemical Identifiers
| Black | |
| Non-sterile | |
| Reusable |
| Phenolic Resin | |
| Pulp/Vinyl | |
| Not Autoclavable |
Specifications
| Black | |
| 576 Cs. | |
| Bagged | |
| Pulp/Vinyl | |
| Reusable | |
| Wide mouth bottle |
| Phenolic Resin | |
| 12/Pk. | |
| Non-sterile | |
| 22-400 | |
| Lined Cap (Smooth) | |
| Not Autoclavable |