missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Black 6000 Series Thermal Transfer Printer Ribbons

Designed for clean room applications. Brady™ Black 6000 Series Thermal Transfer Printer Ribbons have ribbon core inside diameter of 1 in. (25.40 mm), ink side out.

$50.43 - $281.09

Specifications

Product Type Printer Ribbon
Color Black
For Use With (Equipment) Brady Label Printers
Material Polyester
Length (English) 984 ft.
View More Specs

Products 3
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
19104625
View Documents Promotion Details
Brady™
R6002PC
3.27 in.
Each for $217.13
Only null left
 
19104631
View Documents Promotion Details
Brady™
R6007PC
4.33 in.
Each for $281.09
Only null left
 
19104634
View Documents Promotion Details
Brady™
BRDR6011
0.8 in.
Each for $50.43
Only null left
 
Description

Description

  • Plastic ribbon core
  • Resin formulation
Specifications

Specifications

Printer Ribbon
Brady Label Printers
984 ft.
Black
Polyester
Videos
SDS
Documents

Documents