missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Black 6000 Series Thermal Transfer Printer Ribbons
Designed for clean room applications. Brady™ Black 6000 Series Thermal Transfer Printer Ribbons have ribbon core inside diameter of 1 in. (25.40 mm), ink side out.
Supplier: Brady™ R6002PC
Description
- Plastic ribbon core
- Resin formulation
Chemical Identifiers
| Printer Ribbon | |
| Brady Label Printers | |
| 984 ft. |
| Black | |
| Polyester |
Specifications
| Printer Ribbon | |
| Brady Label Printers | |
| 984 ft. |
| Black | |
| Polyester | |
| 3.27 in. |