missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-Biosciences BL21 (DE3) Electro Competent Cells
Description
It is deficient in proteases Lon and OmpT, resistance to phage T1 (fhuA). It is suitable for protein expression under T7 promoter.
Specifications
Specifications
| Product Type | Competent Cells |
| Sterility | Sterile |
| Species | E. coli |
| Format | Vial |
| For Use With (Application) | DNA Transformation |
| Quantity | 6 x 100 μL |