missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biuret Reagent TS, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Biuret Reagent |
| Color | Blue |
| Name Note | Test Solution |
| CAS | 7732-18-5, 1310-73-2, 6381-59-5, 7758-99-8 |
| CAS Min % | 91.0, 6.22, 0.5, 0.13 |
| CAS Max % | 95.0, 6.48, 0.52, 0.13 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | >12 |
| Quantity | 100 mL |
| Show More |