missing translation for 'onlineSavingsMsg'
Learn More

Biuret Reagent, For Serum Protein, LabChem™

Catalog No. LC116901
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC116901 500 mL
LC116902 1 L
LC116904 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC116901 Supplier LabChem Supplier No. LC116901

Specifications

Chemical Name or Material Biuret Reagent
Color Blue
Name Note For Serum Protein
CAS Max % 99.24, 0.6, 0.15, 0.01
CAS 6381-59-5, 7758-99-8, 1310-73-2
Linear Formula CuSO4·5H2O
Solubility Information Soluble in water
Formula Weight 249.68
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sulfur compounds; Fume; Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Show More Show Less