missing translation for 'onlineSavingsMsg'
Learn More

Biuret Reagent, Cutter Lab Formulation, Ricca Chemical

Catalog No. R10225
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R10225 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R10225 Supplier Ricca Chemical Company Supplier No. 10225
Only null left

Specifications

Chemical Name or Material Biuret Reagent
Color Blue
Name Note Cutter Lab Formulation
CAS 7732-18-5, 6132-04-3, 497-19-8, 7758-99-8
CAS Min % 76.0, 13.15, 7.6, 1.31
CAS Max % 79.0, 13.68, 7.91, 1.37
Grade Laboratory
Packaging Cubitainer
pH ∼10.6
Quantity 20 L
Physical Form Liquid
Show More Show Less