missing translation for 'onlineSavingsMsg'
Learn More

Biuret (contains ca. 10% Triuret) 98.0+%, TCI America™

Supplier:  TCI America B0513500G

 View more versions of this product

Catalog No. B0513500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Biuret (contains ca. 10% Triuret)
193°C
C2H5N3O2
500 g
OHJMTUPIZMNBFR-UHFFFAOYSA-N
carbamoylurea
7913
103.08
Crystalline Powder
108-19-0
White
MFCD00007946
biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
C(=O)(N)NC(=O)N
103.081
CHEBI:18138
≥98.0% (N)

Chemical Identifiers

108-19-0
103.081
OHJMTUPIZMNBFR-UHFFFAOYSA-N
7913
carbamoylurea
C2H5N3O2
MFCD00007946
biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
CHEBI:18138
C(=O)(N)NC(=O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions