missing translation for 'onlineSavingsMsg'
Learn More

Biuret (contains ca. 10% Triuret) 98.0+%, TCI America™

Catalog No. p-7118490
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B051325G 25 g
B0513500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B051325G Supplier TCI America Supplier No. B051325G

Chemical Identifiers

CAS 108-19-0
Molecular Formula C2H5N3O2
Molecular Weight (g/mol) 103.081
MDL Number MFCD00007946
InChI Key OHJMTUPIZMNBFR-UHFFFAOYSA-N
Synonym biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
PubChem CID 7913
ChEBI CHEBI:18138
IUPAC Name carbamoylurea
SMILES C(=O)(N)NC(=O)N

Specifications

Melting Point 193°C
Color White
Quantity 25 g
Formula Weight 103.08
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Biuret (contains ca. 10% Triuret)