missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ BiTek™ Fermentation Broth
GREENER_CHOICE

Catalog No. 259520
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
259520 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 259520 Supplier Gibco™ Supplier No. 259520

A dehydrated culture media and supplement product for microbiological testing.

Gibco™ BiTek™ Fermentation Broth is a bacterial fermentation formulated with ingredients that exhibit superior consistency, performance, and solubility.

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Bacterial fermentation
Product Line BiTek
Quantity 10 kg