missing translation for 'onlineSavingsMsg'
Learn More

Bistrifluoroacetamide 98.0+%, TCI America™

Catalog No. p-7118783
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09865G 5 g
B098625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09865G Supplier TCI America Supplier No. B09865G
Only null left

Chemical Identifiers

CAS 407-24-9
Molecular Formula C4HF6NO2
Molecular Weight (g/mol) 209.05
MDL Number MFCD00013564
InChI Key GMQVFHZSXKJCIV-UHFFFAOYSA-N
Synonym BTFA
PubChem CID 67885
IUPAC Name 2,2,2-trifluoro-N-(2,2,2-trifluoroacetyl)acetamide
SMILES FC(F)(F)C(=O)NC(=O)C(F)(F)F

Specifications

Melting Point 84°C
Color White
Boiling Point 120°C
Quantity 5 g
Formula Weight 209.05
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Bistrifluoroacetamide