accessibility menu, dialog, popup

UserName

CAS RN 407-24-9

IUPAC Name: 2,2,2-trifluoro-N-(2,2,2-trifluoroacetyl)acetamide
Synonyms: 2,2,2-trifluoro-N-(2,2,2-trifluoroacetyl)acetamide
Molecular Weight (g/mol): 209.05
Molecular Formula: C4HF6NO2
InChi Key: GMQVFHZSXKJCIV-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)NC(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results