missing translation for 'onlineSavingsMsg'
Learn More

BisphenolF bis(3-chloro-2-hydroxypropyl) ether Qualitative Standard, ≥90% (Mixture of Isomers, HPLC), MilliporeSigma™ Supelco™

Catalog No. 111004207
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004207 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004207 Supplier Merck Emd Millipore Supplier No. 15139250MG

Bisphenol F bis(3-chloro-2-hydroxypropyl) ether a additive used during synthesis of organosol resin preventing theremal degradation of polymer. During the removal of hydrochloric acid in organosol resin results in this chlorinated compound.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Physical Form Neat
Linear Formula C19H22 Cl2O4
MDL Number MFCD01862949
Synonym Bis[4-(3-chloro-2-hydroxypropoxy)phenyl]methane
Type Standard
Quantity 250 mg
Formula Weight 385.28
Percent Purity ≥90% (mixture of isomers, HPLC)
Grade Qualitative Standard