missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol M, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019901
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019901 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019901 Supplier Merck Emd Millipore Supplier No. 23454100MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 13595-25-0
Physical Form Neat
Linear Formula C6H4 [C(CH3)2C6H4 OH]2
MDL Number MFCD00134688
Melting Point 135°C to 139°C
Synonym 4,4 ′-(1,3-Phenylenediisopropylidene)bisphenol; Bisphenol M
Type Analytical Standard
Quantity 100 mg
Formula Weight 346.46
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Show More Show Less