missing translation for 'onlineSavingsMsg'
Learn More

BisphenolA bis(3-chloro-2-hydroxypropyl) ether, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014270
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014270 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014270 Supplier Merck Emd Millipore Supplier No. 15136250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 4809-35-2
Physical Form Neat
Linear Formula C21H26 Cl2O4
MDL Number MFCD01862947
Synonym 2,2-Bis[4-(3-chloro-2-hydroxypropoxy)phenyl]propane; BADGE-2 HCl
Type Analytical Standard
Quantity 250 mg
Formula Weight 413.33
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard