missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol FL, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019904
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019904 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019904 Supplier Merck Emd Millipore Supplier No. 97009100MG
Only null left
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 3236-71-3
Physical Form Neat
Linear Formula C25H18O2
MDL Number MFCD00191392
Melting Point 224°C to 226°C
Synonym 4,4′-(9-Fluorenylidene)diphenol; 9,9-Bis(4-hydroxyphenyl)fluorene; BPFL; BisP-FL
Type Analytical Standard
Quantity 100 mg
Formula Weight 350.41
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Show More Show Less