missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisphenol BP, ≥98% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 1844-01-5 |
| Linear Formula | C25H20O2 |
| MDL Number | MFCD00045785 |
| Synonym | 1,1-Bis(4-hydroxyphenyl)-1,1-diphenylmethane; 4,4′-(Diphenylmethylene)diphenol; 4,4′-Dihydroxytetraphenylmethane; Bis(4-hydroxyphenyl)diphenylmethane; NSC 30848 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 352.43 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Analytical Standard |